Learn More
Medchemexpress LLC Carbamic acid, N,N-dimethyl-, 3-[[[4-(aminomethyl)-1-(5-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-yl)-4-piperidinyl]carbonyl]amino]phenyl ester | 1192189-69-7 | 99.5% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY1265910MM/1ML
Encompass_Preferred
LX7101 is a potent inhibitor of LIM-kinase, ROCK, and PKA. It exhibits significant selectivity for LIMK2, with IC50 values of 4.3 nM for LIMK2 and 32 nM for LIMK1 at 2 μM ATP. This compound shows potential for research in ocular hypertension and associated glaucoma.
  • Potent LIM-kinase, ROCK, and PKA inhibitor
  • Shows significant selectivity for LIMK2
  • Has potential for ocular hypertension research
  • Has potential for associated glaucoma research
  • Reduces intraocular pressure in ocular normotensive rabbits
Catalog No. 50-003-42073