Learn More
Medchemexpress LLC Carbamodithioic acid, N,N-diethyl-, sodium salt (1:1) | 148-18-5 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB1637
Encompass_Preferred
Ditiocarb sodium (Sodium diethyldithiocarbamate) is an orally active copper reagent. It exhibits various activities including antioxidation, chelation, anti-tumor effects, immunomodulation, and anti-HIV properties. This compound is suitable for research in tumors, inflammatory, and immune-related diseases.
  • Orally active copper reagent
  • Exhibits antioxidation properties
  • Shows chelation capabilities
  • Has anti-tumor effects
  • Provides immunomodulation
  • Demonstrates anti-HIV properties
  • Useful for research of tumors
  • Applicable for inflammatory disease research
  • Suitable for immune-related disease research
Catalog No. 50-004-14082