Learn More
Medchemexpress LLC Carbamodithioic acid, N,N-diethyl-, sodium salt (1:1) | 148-18-5 | 99.7% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1637500MG
Encompass_Preferred
Ditiocarb sodium is an orally active copper reagent with diverse biological activities, including antioxidation, chelation, anti-tumor effects, immunomodulation, and anti-HIV properties. It is utilized in the research of tumors, inflammatory, and immune-related diseases. This compound accelerates the cementation of copper from copper sulfate solution on zinc and has been shown to inhibit Cisplatin-induced nephrotoxicity and gastrointestinal toxicity in rats, as well as exhibiting antitumor activity in mouse skin tumor models.
  • Exhibits antioxidation, chelation, anti-tumor, immunomodulation, and anti-HIV properties
  • Useful in research for tumors, inflammatory, and immune-related diseases
  • Accelerates copper cementation from copper sulfate solution
  • Inhibits Cisplatin-induced nephrotoxicity and gastrointestinal toxicity
  • Demonstrates antitumor activity in mouse skin tumor models
Catalog No. 50-004-11845