Learn More
Cayman Chemical Carbaprostacycln mthyl St 10mg
SDP
Supplier:  Cayman Chemical 900018310 MG
Encompass_Preferred
A more lipid soluble form of carbaprostacyclin; may more readily enter cells to activate PPARδ; may be more amenable for dietary formulations
Catalog No. 50-289-3547