Learn More
Cayman Chemical Carbaslate CalcIum 500mg
SDP
Supplier:  Cayman Chemical 29806500 MG
Encompass_Preferred
A calcium-urea chelate of acetylsalicylate; has analgesic, anti-inflammatory, and antipyretic effects; reduces carrageenan-induced paw edema in rats
Catalog No. 50-299-0032