Learn More
Apexbio Technology LLC Carbazochrome sodium sulfonate (AC-17) 51460-26-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19035.1
Encompass_Preferred
Carbazochrome sodium sulfonate (AC-17 CAS 51460-26-5) is a small molecule frequently utilized for investigating mechanisms underlying capillary stability and hemostasis It acts primarily by stabilizing endothelial structures within capillaries enhancing platelet aggregation and decreasing vascular permeability consequently minimizing microvascular leakage and bleeding In experimental settings AC-17 serves as a tool to elucidate pathways involved in vascular permeability regulation and platelet-mediated hemostatic responses using both in vitro and in vivo models It is particularly valuable for pharmacological studies related to capillary dysfunction and hemorrhagic disorders
Catalog No. 50-000-61990