Learn More
Cayman Chemical CarbazomycIn C 5mg
SDP
Supplier:  Cayman Chemical 330905 MG
Encompass_Preferred
A bacterial metabolite with diverse biological activities; active against S. aureus, B. anthracis, B. subtilis, and M. flavus (MICs = 50, 25, 100, and 50 UG/ml, respectively), the fungi T. asteroides and T. mentagrophytes (MICs = 25 and 100 UG/ml, respectively), and P. falciparum (IC50 = 2.1 UG/ml); active against a panel of five plant pathogenic fungi (MICs = 12.5-100 UG/ml); cytotoxic to MCF-7, KB, and NCI H187 cells (IC50s = 9.8, 21.4, and 8.2 UG/ml, respectively); inhibits 5-LO activity in RBL-1 cell extracts (IC50 = 1.9 µM)
Catalog No. 50-294-7625