Learn More
Apexbio Technology LLC Carbenicillin(Synonyms: Pyopen, Carbenicillin Sodium, Alpha-Carboxybenzylpenicillin, Carboxybenzylpenicillin), 5g, CAS: B3412.
SDP
Supplier:  Apexbio Technology LLC B34125000
Encompass_Preferred
Carbenicillin (CAS 4697-36-3) is a semi-synthetic penicillin derivative characterized by a broad-spectrum antibacterial profile Its bactericidal activity arises primarily through inhibition of bacterial cell wall biosynthesis specifically by interfering with penicillin-binding proteins (PBPs) thus impairing peptidoglycan formation This compound is widely utilized in biomedical research laboratories particularly for selecting bacteria carrying plasmids encoding carbenicillin resistance markers Carbenicillin enables effective differentiation between plasmid-containing and non-plasmid-containing bacterial colonies in molecular biology studies
Catalog No. 50-000-60441