Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbenicillin disodium | 4800-94-6 | 98.12% | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYB0525A

Encompass_Preferred

Carbenicillin disodium is a broad-spectrum, semi-synthetic penicillin antibiotic effective against gram-negative bacteria. It works by interfering with cell wall synthesis and demonstrates low toxicity to plant tissue.

  • Broad-spectrum semi-synthetic penicillin antibiotic
  • Effective against gram-negative bacteria
  • Interferes with cell wall synthesis
  • Low toxicity to plant tissue
  • Unstable in solutions; fresh preparation recommended
  • Store at 4°C, sealed, away from moisture and light
  • Purity of 98.12%
  • Soluble in H2O at 50 mg/mL

Catalog No. 50-004-05803


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.