Learn More
Selleck Chemical LLC Carbenicillin disodium 50mg 4800-94-6 BRL-2064
SDP
Supplier:  Selleck Chemical LLC S317950MG
Encompass_Preferred
Carbenicillin (BRL-2064) is a semi-synthetic penicillin antibiotic which interferes with cell wall synthesis of gram-negative bacteria while displaying low toxicity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3465