Learn More
Cayman Chemical CarbetapentncItrate 500mg
SDP
Supplier:  Cayman Chemical 18606500 MG
Encompass_Preferred
A σ1 receptor agonist (Ki = 10.4 nM); inhibits citric acid-induced cough in guinea pigs from 1-5 mg/kg; reduces the number of seizures, mortality, and hippocampal neuronal cell death in a rat model of kainate-induced seizures at 12.5 and 25 mg/kg
Catalog No. 50-296-5942