Learn More
Cayman Chemical CarbetocInacetate 10mg
SDP
Supplier:  Cayman Chemical 3323410 MG
Encompass_Preferred
A peptide oxytocin receptor agonist; binds to oxytocin receptors (Kd = 1.96 nM); selectively binds to oxytocin receptors over vasopressin V2 receptors (Kds = 1.96 and 61.3 nM, respectively); induces contraction of isolated rat uterine strips (EC50 = 48 nM); inhibits oxytocin-induced contraction of isolated rat uterine strips (pA2 = 8.21); increases the frequency and amplitude of uterine contractions in EArly postpartum dairy cows at 0.35 mg/animal; prevents stress-induced reinstatement of morphine-seeking behavior in mice at 6.4 mg/kg; reverses learned helplessness and decreases social withdrawal induced by chronic unpredictable stress in tree shrews
Catalog No. 50-294-6543