Learn More
Electron Microscopy Sciences Carbide Micro Tool Set 1.0 mm
SDP
Supplier:  Electron Microscopy Sciences 6209510
Encompass
These unique micro cutting tools are made using the latest grinding and polishing technology producing the highest quality tool on the market today. Each set comes with a MT-1 Handle, and instrument case. Tips are Micro-Needle, Micro-Graver, 3-sided, Micro-Chisel, Micro-Spade, Micro-Knife 20 degree , Micro-Knife 45 degree ,Micro-Graver Oval, and Micro-Scraper
Tip: 1.0 mm (.040")
Catalog No. 50-996-699