Learn More
Selleck Chemical LLC Carbidopa 25mg 28860-95-9
SDP
Supplier:  Selleck Chemical LLC S189125MG
Encompass_Preferred
Carbidopa is an aromatic-L-amino-acid decarboxylase inhibitor with an IC50 of 29 ¿ 2 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8198