Learn More
Medchemexpress LLC Carbidopa | 28860-95-9 | 99.8% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYB031110MM/1ML
Encompass_Preferred
Carbidopa, also known as (S)-(-)-Carbidopa, is a peripheral decarboxylase inhibitor primarily used for research related to Parkinson's disease. It also functions as a selective aryl hydrocarbon receptor (AhR) modulator and has been shown to inhibit pancreatic cancer cell and tumor growth.
  • Acts as a peripheral decarboxylase inhibitor
  • Functions as a selective aryl hydrocarbon receptor (AhR) modulator
  • Demonstrates anticancer activity by inhibiting pancreatic cancer cell and tumor growth
  • High purity of 99.84% ensures reliability for research applications
  • Comprehensive documentation available, including Data Sheet, COA, SDS, HNMR, RP-HPLC, and Handling Instructions
  • Detailed solubility information provided for in vitro and in vivo applications, with protocols for preparing clear stock and working solutions
  • Intended for research use only
Catalog No. 50-004-06729