Learn More
Medchemexpress LLC Carbidopa | 28860-95-9 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0311100MG
Encompass_Preferred
Carbidopa is a peripheral decarboxylase inhibitor used in Parkinson's disease research. It acts as a selective aryl hydrocarbon receptor (AhR) modulator and has shown to inhibit pancreatic cancer cell and tumor growth.
  • Peripheral decarboxylase inhibitor
  • Used in Parkinson's disease research
  • Selective aryl hydrocarbon receptor (AhR) modulator
  • Inhibits pancreatic cancer cell and tumor growth
Catalog No. 50-004-05854