Learn More
Selleck Chemical LLC Carbidopa S1891-100mg
SDP
Supplier:  Selleck Chemical LLC S1891100MG
Encompass_Preferred
Carbidopa is an aromatic-L-amino-acid decarboxylase inhibitor with an IC50 of 29 2 M
Catalog No. 50-311-0759