Learn More
Medchemexpress LLC Carbidopa ((S)-(-)-Carbidopa) | 28860-95-9 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB031150MG
Encompass_Preferred
Carbidopa ((S)-(-)-Carbidopa) is a peripheral decarboxylase inhibitor that can be utilized for Parkinson's disease research. It acts as a selective aryl hydrocarbon receptor (AhR) modulator and has been shown to inhibit pancreatic cancer cell and tumor growth.
  • Peripheral decarboxylase inhibitor.
  • Used in Parkinson's disease research.
  • Selective aryl hydrocarbon receptor (AhR) modulator.
  • Inhibits pancreatic cancer cell and tumor growth.
Catalog No. 50-004-05466