Learn More
Medchemexpress LLC HY-B0558 500mg Medchemexpress, Carbimazole CAS:22232-54-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0558500MG
Encompass_Preferred
Medchemexpress, HY-B0558 500mg Carbimazole CAS:22232-54-8 Carbimazole is an imidazole antithyroid agent and can be used for the research of Graves' disease. Carbimazole plays its role due to its rapid conversion to methylmercapto imidazole (MMI) in vivo and can be converted to methimazole in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2599