Learn More
Medchemexpress LLC HY-B1589A 100mg Medchemexpress, Carbinoxamine maleate salt CAS:3505-38-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1589A100MG
Encompass_Preferred
Medchemexpress, HY-B1589A 100mg Carbinoxamine maleate salt CAS:3505-38-2 Carbinoxamine maleate salt is a histamine H1 receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2973