Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbinoxamine maleate salt | 3505-38-2 | 98.78% | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYB1589A

Encompass_Preferred

Carbinoxamine maleate salt is a histamine H1 receptor antagonist available as a white to off-white solid. It has a high purity of 98.78% and a molecular formula of C20H23ClN2O5.

  • Targets histamine H1 receptor
  • 98.78% purity
  • Molecular weight of 406.86
  • White to off-white solid appearance
  • Recommended storage at 4°C, sealed and away from moisture

Catalog No. 50-004-14253


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.