Learn More
Medchemexpress LLC Carbocisteine | 638-23-3 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYD0205A1G
Encompass_Preferred
Carbocisteine is an orally active mucolytic agent. It attenuates the phosphorylation of NF-κB p65 and ERK1/2, modulates Nrf2/HO-1 and NFκB interplay, and inhibits apoptosis. It is used in chronic obstructive pulmonary disease (COPD) research.
  • Orally active mucolytic agent
  • Attenuates the phosphorylation of NF-κB p65 and ERK1/2
  • Modulates Nrf2/HO-1 and NFκB interplay
  • Inhibits apoptosis
  • Used in chronic obstructive pulmonary disease (COPD) research
Catalog No. 50-004-10226