Learn More
Medchemexpress LLC Carbocysteine sulfoxide | 5439-87-2 | MFCD21364079 | 99.0% | 195.19 g/mol | C5H9NO5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW42236210MG
Encompass_Preferred
Carbocysteine sulfoxide is a small-molecule impurity of carbocisteine intended for research use. The substance is identified by CAS number 5439-87-2, has the molecular formula C5H9NO5S, and a molecular weight of 195.19 g/mol. It is provided in small-mass packages for laboratory and analytical applications.
  • Used as an impurity reference for analytical method development.
  • Supplied in small-mass packages suitable for laboratory research.
  • Identified by CAS number 5439-87-2 for substance tracking.
  • Molecular formula C5H9NO5S and molecular weight 195.19 g/mol.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-81564