Learn More
Cayman Chemical Carbofurn-d3 1mg
SDP
Supplier:  Cayman Chemical 395581 MG
Encompass_Preferred
An internal standard for the quantification of carbofuran by GC-or LC-MS
Catalog No. 50-291-8865