Learn More
Waters Corp Waters Carbohydrate Analysis Column, Amino (NH2), 125 Å, 10 μm
Supplier:  Waters Corp WAT084038
Encompass_Preferred
This Carbohydrate Analysis Column features covalently bonded amino packing, ideal for separating low molecular weight sugars (mono-, di-, and trisaccharides). It provides high-resolution carbohydrate profiling in complex sample matrices. Optimized for 65-85% acetonitrile in water mobile phase and ambient to 35°C temperatures, its 125 Å pore size and 10 μm particle size enable detailed sugar analysis. The robust amino-based stationary phase offers strong retention and selectivity.
  • Covalently bonded amino packing
  • Separates low molecular weight sugars (mono-, di-, and trisaccharides)
  • Optimized for 65-85% acetonitrile in water mobile phase
  • Reliable, reproducible separations up to 35°C
  • Robust amino-based stationary phase offers strong retention and selectivity
  • Integrates into LC-MS workflows for carbohydrate analysis
Catalog No. 50-785-901