Learn More
Medchemexpress LLC Carbomer | 9007-20-9 | MFCD00146299 | 95.0% | 1,250,000 g/mol (approx.) | Poly(acrylic acid) (C3H4O2)n | 50 G
SDP
Supplier:  Medchemexpress LLC HYW250721C50G
Encompass_Preferred
Carbomer 941 is a crosslinked poly(acrylic acid) polymer used as a thickening and gelling agent in pharmaceutical and laboratory formulations. It forms high-viscosity, stable gels suitable for topical preparations, controlled-release systems, and other research applications. Supplied for research use with accompanying COA and SDS documentation.
  • Forms high-viscosity, stable gels
  • Functions as a thickening and suspending agent
  • Suitable for topical and controlled-release formulations
  • Low residual acrylic acid and low loss on drying per COA
  • Provided with COA and SDS for quality and safety verification
Catalog No. 50-002-81437