Learn More
Medchemexpress LLC Carbomer 940 | 9007-20-9 | 95.0% | Not specified (crosslinked polymer; no defined molecular weight) | (C3H4O2)n | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW250721A100MG
Encompass_Preferred
Carbomer 940 is a crosslinked polyacrylic acid polymer used as a high-efficiency thickening and gelling agent for pharmaceutical, cosmetic, and biochemical applications. It appears as a white to off-white powder and forms stable gels that control viscosity and enable topical and local drug delivery, suspension stabilization, and controlled-release formulations.
  • Provides high viscosity and gel-forming properties.
  • Forms a stable gel matrix for controlled drug release.
  • Resistant to heat, light, and microbial contamination.
  • Suitable for topical and local formulations.
  • Purity typically 95.0% for research-grade material.
Catalog No. 50-002-81206