Learn More
Medchemexpress LLC Carbomer 941 | 9007-20-9 | MFCD00084394 | 95.0% | ≈1,250,000 g/mol | (C3H4O2)n | 500 G
SDP
Supplier:  Medchemexpress LLC HYW250721C500G
Encompass_Preferred
Carbomer 941 is a crosslinked poly(acrylic acid) high-molecular-weight polymer used as a thickening, gelling, and stabilizing agent in pharmaceutical and formulation applications. It forms high-viscosity gels and provides a stable matrix for topical formulations and controlled drug release; it is typically supplied as a dry powder and stored at room temperature, protected from moisture.
  • High viscosity gel formation suitable for topical formulations.
  • Stable gel matrix for controlled drug release.
  • Effective thickening at low concentrations.
  • Good resistance to heat, light, and microbial contamination when formulated properly.
  • Compatible with common solvents and excipients used in formulations.
  • Available in multiple package sizes for laboratory and production use.
Catalog No. 50-002-81432