Learn More
Medchemexpress LLC Carbomer 941 (CBM 941) | 9007-20-9 | MFCD01675509 | 95.0% | ≈1,250,000 g/mol | C42H80O8 | 250 G
SDP
Supplier:  Medchemexpress LLC HYW250721C250G
Encompass_Preferred
Carbomer 941 is a crosslinked acrylic acid polymer used as a rheology modifier and thickening agent in formulation and pharmaceutical research. It forms high-viscosity, stable gels and provides a robust gel matrix for controlled release and topical applications. Supplied as a white to off-white powder with documented purity for laboratory use.
  • Forms high-viscosity, stable gels.
  • Acts as a rheology modifier and thickening agent.
  • Provides controlled release and gel matrix stability.
  • White to off-white solid powder, easy to weigh and disperse.
  • Compatible with aqueous formulations and common excipients.
  • Supplied with purity specification for quality control.
Catalog No. 50-002-80511