Learn More
Sigma Aldrich Fine Chemicals Biosciences Carbon disulfide ACS reage
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1801732L
Encompass
Carbon disulfide is a highly volatile flammable liquid with low ignition temperature. It can be synthesized by reacting hydrocarbon gas with sulfur. It is an important raw material for preparing viscose rayon and cellophane film.
Catalog No. 50-289-9292