Learn More
TELEDYNE CETAC TECHNOLOGIES Sample Probes for XLR-8 Auto Sampler, 0.5 OD, Blue
SDP
Supplier:  TELEDYNE CETAC TECHNOLOGIES SP5800C/DEL
Encompass
Length: 10 inches (25.4 cm), Probe Material: FEP-covered carbon fiber, Capillary Material: PFA, Capillary Length: 108 in (2.74 m)
Catalog No. NC0150258