Learn More
Leica Biosystems CARBON FILTER FOR ASP 300
Supplier:  Leica Biosystems 14047634150
Encompass_Preferred
  • Compatible with Leica ASP300 and ASP300S automated tissue processors.
  • Activated carbon filter type for effective vapor filtration.
  • Designed for histology laboratories to maintain air quality.
  • Absorbs hazardous organic and inorganic vapors, including xylene, formalin, methanol, toluene, and isopropyl alcohol.
  • Recommended replacement interval of 3 months.
  • Length of filter is 11.25 in.
  • Width of filter is 5.75 in.
  • Height of filter is 0.75 in.
  • Manufactured by Leica Microsystems Inc.
Catalog No. NC0446459