Learn More
Ted Pella Inc Carbon Tape, Aluminum Base 8mm
SDP
Supplier:  Ted Pella Inc 160868
Encompass
Smooth surface superior to traditional woven carbon tape for SEM and EDS analysis of small fibers and powders. Ideal for automated analysis of particles. Width: 8mm
Catalog No. NC1784524