Learn More
Bioworld Carbonate-Bicarbonate Buffer Packs, 0.2M, 40 PK(s)
Supplier:  Bioworld 402200921
Encompass_Preferred
Product DescriptionEach pack yields 0.2M carbonate-bicarbonate buffer and has a pH at 9.4 when dissolved in 500 ml deionized water, 40 packs per order. These packs are ideal for microplate coating for RIA and EIA techniques. Dry-blend packs of carbonate-bicarbonate buffer are easy to use. Simply empty contents of one foil envelope pack into a beaker, add ultrapure water and stir to dissolve. The packs eliminate weighing time and pH adjustments.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-481