Learn More
YSI 3P LifeScience 1579 Carbonate Buffer Concentrate
Supplier:  YSI 3P LifeScience 1579
Encompass
Carbonate buffer concentrate for use with 2725 Methanol Membranes and 2786 Ethanol Membranes. Reconstitute to make 1 liter.
Catalog No. NC9799965