Learn More
Medchemexpress LLC Carbonic acid, 1-[4-[(16-azido-PEG3)oxy]-5-methoxy-2-nitrophenyl]ethyl 2,5-dioxo-pyrrolidinyl ester | 1802907-96-5 | MFCD29052189 | 98.7% | 640.60 g/mol | C26H36N6O13 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW19093750MG
Encompass_Preferred
PC Azido-PEG3-NHS carbonate ester is a photocleavable azido-PEG3 NHS carbonate linker for labeling primary amines on proteins, amine-modified oligonucleotides, and other amine-containing molecules. It combines an NHS carbonate reactive group with an azide handle and a photocleavable moiety to enable covalent labeling and light-triggered release. Supplied as a colorless to light-yellow oil for research use; follow manufacturer storage recommendations.
Catalog No. 50-002-80727