Learn More
Medchemexpress LLC Carbonic Anhydrase 2 10ug
SDP
Supplier:  Medchemexpress LLC HYP7286110UG
Encompass_Preferred
Carbonic Anhydrase 2 Protein is a member of the carbonic anhydrase isozyme family responsible for catalyzing the reversible hydration of carbon dioxide Mutations in CA2 are linked to osteopetrosis and renal tubular acidosis The gene exhibits biased expression in various tissues notably in the stomach and colon CA2 plays vital role in the regulation of ion transport and pH balance and is involved in many biological processes Carbonic Anhydrase 2 Protein Mouse (HEK293 His) is the recombinant mouse-derived Carbonic Anhydrase 2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81586