Learn More
Medchemexpress LLC Carbonic Anhydrase 2 5ug
SDP
Supplier:  Medchemexpress LLC HYP728605UG
Encompass_Preferred
Carbonic Anhydrase 2 (CA2) catalyzes the reversible hydration of carbon dioxide Mutations in CA2 are linked to osteopetrosis and renal tubular acidosis The gene exhibits biased expression in various tissues notably in the stomach and colon Two transcript variants encoding different isoforms have been identified Carbonic Anhydrase 2 Protein Human (C-His Solution) is the recombinant human-derived Carbonic Anhydrase 2 protein expressed by E coli with C-His labeled tag
Catalog No. 50-001-81892