Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbonic Anhydrase 2 Antibody (YA3362) | 50 UL
SDP

Supplier:  Medchemexpress LLC HYP8361750UL

Encompass_Preferred

Carbonic Anhydrase 2 Protein is a member of the carbonic anhydrase isozyme family, responsible for catalyzing the reversible hydration of carbon dioxide. Dysregulation of this enzyme is linked to conditions such as osteopetrosis and renal tubular acidosis. This protein also exhibits biased expression in various tissues, including the stomach and colon.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Conjugation: Non-conjugated
  • Antibody type: Monoclonal IgG antibody
  • Target: Carbonic anhydrase 2
  • Research field: Cardiovascular
  • Formulation: Supplied in PBS, pH 7.4
  • Appearance: Liquid
  • Applications: Western blot (WB), immunohistochemistry-paraffin (IHC-P)
  • Storage stability: Stable for 1 year when stored at -20°C

Catalog No. 50-004-43461


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.