Learn More
Medchemexpress LLC Carbonic Anhydrase 3 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP77195UG
Encompass_Preferred
Carbonic Anhydrase 3 Protein Human (His) expresses in E coli with a His tag at the N-terminus Carbonic anhydrase 3 (CA III) is a zinc metalloenzyme known to catalyze the reversible hydration of CO2 to HCO3-[1]
Catalog No. 50-001-86421