Learn More
Medchemexpress LLC Carbonic Anhydrase 4 50ug
SDP
Supplier:  Medchemexpress LLC HYP7286250UG
Encompass_Preferred
Carbonic anhydrase 4 catalyzes the reversible hydration of carbon dioxide and plays a crucial role in maintaining intracellular and extracellular pH Carbonic Anhydrase 4 Protein Human (HEK293 His) is the recombinant human-derived Carbonic Anhydrase 4 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81627