Learn More
Medchemexpress LLC Carbonic anhydrase 9 protein | >95.0% | 250UG

Supplier: Medchemexpress LLC HYP70064250UG
Carbonic Anhydrase 9 Protein is a recombinant human CA9 expressed in HEK293 cells with a C-terminal 6xHis tag. Supplied as a purified protein (250 μg, >95% purity), it is suitable for biochemical assays, antibody validation, and reagent development. The protein is glycosylated (approx. 45-60 kDa) and should be aliquoted and stored at -20°C or -80°C for extended storage.
- Recombinant human protein expressed in HEK293 cells.
- C-terminal 6xHis tag for detection and purification.
- Greater than 95% purity as determined by reducing SDS-PAGE.
- Glycosylated form, approximately 45-60 kDa.
- Suitable for biochemical assays and antibody validation.
- Stable when aliquoted and stored at -20°C or -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.