Learn More
Medchemexpress LLC Carbonic anhydrase 9 protein, human (HEK293, His) | >95.0% | 500UG

Supplier: Medchemexpress LLC HYP70064500UG
Recombinant human Carbonic Anhydrase 9 (CA9) produced in HEK293 cells with a C-terminal 6xHis tag. CA9 catalyzes the reversible hydration of carbon dioxide to bicarbonate and protons and is commonly used in cancer biology research and biochemical assays. The protein is glycosylated and supplied as a purified preparation for research applications.
- Recombinant human CA9 expressed in HEK293 cells.
- C-terminal 6xHis tag for detection and purification.
- Glycosylated protein; molecular weight approx. 45-60 kDa.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Supplied for research use in pack sizes including 500 μg.
- Suitable for biochemical assays and antibody validation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.