Learn More
Medchemexpress LLC Carbonic Anhydrase 9 50ug
SDP
Supplier:  Medchemexpress LLC HYP7286350UG
Encompass_Preferred
The carbonic anhydrase 9 (CA9) protein plays a crucial role as a catalyst in the conversion of carbon dioxide and water It promotes the formation of bicarbonate and hydrogen ions (dissociated ions of carbonic acid) Carbonic Anhydrase 9 Protein Human (HEK293 Fc) is the recombinant human-derived Carbonic Anhydrase 9 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-83109