Learn More
Medchemexpress LLC Carbonic anhydrase 9 (recombinant protein) | 97.0% | 20 UG
SDP
Supplier:  Medchemexpress LLC HYP7545420UG
Encompass_Preferred
Recombinant canine Carbonic Anhydrase 9 (CA9/CA-IX) produced in HEK293 cells with a C-terminal His tag. Supplied as a purified research reagent with reported purity of 97.0% and an approximate molecular weight of 42-55 kDa; pack size noted as 20 μg from the supplier SKU. Intended for antibody characterization, binding studies, and immunoassay development.
Catalog No. 50-002-93417