Learn More
Medchemexpress LLC Carbonic anhydrase, Bovine erythrocytes | 9001-03-0 | 29.0 kDa | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP17755MG
Encompass_Preferred
Carbonic anhydrase, Bovine erythrocytes (EC 4.2.1.1) is a ubiquitous zinc-containing metalloenzyme present in prokaryotes and eukaryotes. It catalyzes the reversible conversion of carbon dioxide to bicarbonate and protons. This enzyme is primarily used for research applications in areas such as cancer, glaucoma, obesity, and epilepsy.
  • Catalyzes reversible conversion of carbon dioxide to bicarbonate and protons
  • Used in research for cancer, glaucoma, obesity, and epilepsy
  • Zinc-containing metalloenzyme
  • Optimal pH of 7.0-7.5
  • Inhibitors include monovalent anions, sulfonates, sulfonamides, and imidazole
  • Appears as a white to off-white solid
  • Enzyme activity is ≥3000 U/mg solid
  • Soluble in water at concentrations ≥ 50 mg/mL
Catalog No. 50-004-23931