Learn More
Medchemexpress LLC Carbonic Anhydrase I Antibody (YA560) | 29 kDa | 10 UL

Supplier: Medchemexpress LLC HYP8038210UL
Carbonic Anhydrase I Antibody (YA560) is a liquid antibody for research use, non-conjugated and supplied at 1mg/mL concentration. It is intended for laboratory chemicals and the manufacture of other substances.
- Host: Rabbit
- Reactivity: Human, mouse, rat
- Clonality: YA560
- Molecular weight: 29 kDa
- Applications: WB, IHC-P
- Formulation includes 0.05% BSA and 40% Glycerol with 0.05% sodium azide
- Store at -20°C, avoid repeated freeze/thaw cycles
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.