Learn More
Medchemexpress LLC Carbonic anhydrase inhibitor 16 | 4479-70-3 | 98.0% | 302.31 g/mol | C14H10N2O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16310910MG
Encompass_Preferred
Carbonic anhydrase inhibitor 16 is a research-grade small molecule reported to inhibit human carbonic anhydrase isoforms and Dengue protease activity. It is intended for in vitro biochemical assays and exploratory research requiring selective carbonic anhydrase inhibition.
  • Small molecule carbonic anhydrase inhibitor for research use.
  • Potent inhibition of hCA I and hCA II (Ki 28.5 nM and 2.2 nM).
  • Reported CAS number 4479-70-3 for substance identification.
  • Molecular formula C14H10N2O4S; molecular weight 302.31 g/mol.
  • Supplied quantity 10 MG with reported purity 98.0%.
  • Intended for in vitro biochemical assays and pharmacological research.
Catalog No. 50-002-88902