Learn More
Medchemexpress LLC Carbonic anhydrase inhibitor 2 | 2758231-43-3 | 98.4% | 344.34 g/mol | C12H16N4O6S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1428495MG
Encompass_Preferred
Carbonic anhydrase inhibitor 2 is a small-molecule inhibitor of carbonic anhydrase II used for research on enzyme inhibition and ocular pressure modulation. Described in literature as compound 7c, it is supplied as a white to off-white solid (CAS 2758231-43-3) with molecular formula C12H16N4O6S and molecular weight 344.34 g/mol.
  • Potent carbonic anhydrase II inhibitory activity, suitable for biochemical assays.
  • Shown to reduce intraocular pressure in preclinical glaucoma models.
  • High purity: 98.4%, appropriate for research applications.
  • Supplied as a solid, facilitating storage and handling.
  • Available in multiple milligram pack sizes for small-scale studies.
Catalog No. 50-002-21369