Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbonic anhydrase inhibitor 2 | 2758231-43-3 | 98.4% | C12H16N4O6S | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY142849

Encompass_Preferred

Carbonic anhydrase inhibitor 2 is described as a carbonic anhydrase II inhibitor. It has been shown to reduce intraocular pressure in glaucomatous rabbits.

  • Target is CA II
  • Soluble in DMSO (20 mg/mL)
  • In vivo solubility protocols available with DMSO, PEG300, Tween-80, Saline, SBE-β-CD, or Corn Oil

Catalog No. 50-003-56854


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.